b-Amino-3-chlorobenzeneethanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-(3-chlorophenyl)ethanol

OR452168-1G OR452168-250MG
APOSOR452168-1GEA 161 EUR
APOSOR452168-1G APOSOR452168-250MG
b-Amino-3-chlorobenzeneethanol 97%
b-Amino-3-chlorobenzeneethanol
Formel: C₈H₁₀ClNO
Molecular Weight: 171,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827190
CAS-Nummer: 179811-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR