b-Amino-2-chlorobenzeneethanol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-2-(2-chlorophenyl)ethanol
OR452169-1G
APOSOR452169-1GEA
190
EUR
APOSOR452169-1G
b-Amino-2-chlorobenzeneethanol 97%
b-Amino-2-chlorobenzeneethanol
Formel:
C₈H₁₀ClNO Molecular Weight: 171,624 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09750965 CAS-Nummer: 127428-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...