b-Amino-2-bromobenzeneethanol 97%

Lieferant: Apollo Scientific

OR452170-1G OR452170-250MG OR452170-500MG
APOSOR452170-1GEA 783 EUR
APOSOR452170-1G APOSOR452170-250MG APOSOR452170-500MG
b-Amino-2-bromobenzeneethanol 97%
b-Amino-2-bromobenzeneethanol
Formel: C₈H₁₀BrNO
Molecular Weight: 216,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827196
CAS-Nummer: 1184472-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR