b-Amino-4-nitrobenzenepropanol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-(4-nitrophenyl)propan-1-ol
OR452175-1G
OR452175-250MG
APOSOR452175-1GEA
789
EUR
APOSOR452175-1G
APOSOR452175-250MG
b-Amino-4-nitrobenzenepropanol 97%
b-Amino-4-nitrobenzenepropanol
Formel:
C₉H₁₂N₂O₃ Molecular Weight: 196,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17171137 CAS-Nummer: 174302-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...