b-Amino-2-methoxybenzenepropanol 97%
Lieferant: Apollo Scientific
OR452180-1G
OR452180-250MG
OR452180-500MG
APOSOR452180-1GEA
783
EUR
APOSOR452180-1G
APOSOR452180-250MG
APOSOR452180-500MG
b-Amino-2-methoxybenzenepropanol 97%
b-Amino-2-methoxybenzenepropanol
Formel:
C₁₀H₁₅NO₂ Molecular Weight: 181,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10034615 CAS-Nummer: 1018665-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...