(2R)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%
Lieferant: Apollo Scientific
OR452187-1G OR452187-250MG OR452187-500MG
APOSOR452187-1GEA 901 EUR
APOSOR452187-1G APOSOR452187-250MG APOSOR452187-500MG
(2R)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride 97%
(2R)-N1,N1-Dimethyl-1,2-propanediamine dihydrochloride
Formel: C₅H₁₄N₂·2ClH
Molecular Weight: 175,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619104
CAS-Nummer: 403712-75-4