N-Boc-[(2R)-2-amino-3-methylbutyl]amine 97%

Lieferant: Apollo Scientific

OR452194-1G OR452194-250MG OR452194-500MG
APOSOR452194-1GEA 1050 EUR
APOSOR452194-1G APOSOR452194-250MG APOSOR452194-500MG
N-Boc-[(2R)-2-amino-3-methylbutyl]amine 97%
N-Boc-[(2R)-2-amino-3-methylbutyl]amine
Formel: C₁₀H₂₂N₂O₂
Molecular Weight: 202.294 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18677264
CAS-Nummer: 400652-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR