N-Boc-(2-amino-3-methylbutyl)amine 97%
Lieferant: Apollo Scientific
OR452195-1G
OR452195-250MG
OR452195-500MG
APOSOR452195-1GEA
1050
EUR
APOSOR452195-1G
APOSOR452195-250MG
APOSOR452195-500MG
N-Boc-(2-amino-3-methylbutyl)amine 97%
N-Boc-(2-amino-3-methylbutyl)amine
Formel:
C₁₀H₂₂N₂O₂ Molecular Weight: 202.294 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14584430 CAS-Nummer: 1117693-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...