N-Boc-2-amino-butylamine 97%

Lieferant: Apollo Scientific

OR452196-1G OR452196-250MG OR452196-500MG
APOSOR452196-1GEA 654 EUR
APOSOR452196-1G APOSOR452196-250MG APOSOR452196-500MG
N-Boc-2-amino-butylamine 97%
N-Boc-2-amino-butylamine
Formel: C₉H₂₀N₂O₂
Molecular Weight: 188,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660179
CAS-Nummer: 1179533-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR