4-[2-(4-Methylphenyl)ethyl]-piperidine hydrochloride 97%

Lieferant: Apollo Scientific

OR452204-1G OR452204-500MG
APOSOR452204-1GEA 568 EUR
APOSOR452204-1G APOSOR452204-500MG
4-[2-(4-Methylphenyl)ethyl]-piperidine hydrochloride 97%
4-[2-(4-Methylphenyl)ethyl]-piperidine hydrochloride
Formel: C₁₄H₂₁N·ClH
Molecular Weight: 239,784 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28111316
CAS-Nummer: 654662-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR