4-Cyano-4-(3-chlorophenyl)cyclohexanone 97%
Lieferant: Apollo Scientific
OR452208-1G
OR452208-500MG
APOSOR452208-1GEA
609
EUR
APOSOR452208-1G
APOSOR452208-500MG
4-Cyano-4-(3-chlorophenyl)cyclohexanone 97%
4-Cyano-4-(3-chlorophenyl)cyclohexanone
Formel:
C₁₃H₁₂ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11849768 CAS-Nummer: 65619-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...