2-(4-Chlorophenoxy)-benzylamine 97%

Lieferant: Apollo Scientific

OR452224-1G OR452224-500MG
APOSOR452224-1GEA 522 EUR
APOSOR452224-1G APOSOR452224-500MG
2-(4-Chlorophenoxy)-benzylamine 97%
2-(4-Chlorophenoxy)-benzylamine
Formel: C₁₃H₁₂ClNO
Molecular Weight: 233,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11191890
CAS-Nummer: 792158-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR