3-(3-Pyridyl)-L-alanine dihydrochloride
Lieferant: Apollo Scientific
Synonyme:
H-3-(3-Pyridyl)-Ala-OH·2HCl, (S)-2-Amino-3-(pyridin-3-yl)propanoic acid dihydrochloride
OR460016-1G
OR460016-250MG
APOSOR460016-1GEA
369
EUR
APOSOR460016-1G
APOSOR460016-250MG
3-(3-Pyridyl)-L-alanine dihydrochloride
3-(3-Pyridyl)-L-alanine dihydrochloride
Formel:
C₈H₁₀N₂O₂·2ClH Molecular Weight: 239,099 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191178 CAS-Nummer: 93960-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...