1-Bromo-2-iodoethane
Lieferant: Apollo Scientific
|
Achtung
|
OR460045-1G
OR460045-5G
APOSOR460045-1GEA
184
EUR
APOSOR460045-1G
APOSOR460045-5G
1-Bromo-2-iodoethane
1-Bromo-2-iodoethane
Formel:
C₂H₄BrI Molecular Weight: 234,862 g/mol Siedepunkt: 163 °C Schmelzpunkt: 28…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041542 CAS-Nummer: 590-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...