3-Bromo-5-nitroaniline 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-bromonitrobenzene

OR46061-1G OR46061-5G OR46061-25G
APOSOR46061-1GEA 21.9 EUR
APOSOR46061-1G APOSOR46061-5G APOSOR46061-25G
3-Bromo-5-nitroaniline 96%
3-Bromo-5-nitroaniline
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
Schmelzpunkt: 128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00462820
CAS-Nummer: 55215-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR