3-Amino-5-bromophenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-hydroxyaniline

OR46063-1G OR46063-250MG
APOSOR46063-1GEA 357 EUR
APOSOR46063-1G APOSOR46063-250MG
3-Amino-5-bromophenol 97%
3-Amino-5-bromophenol
Formel: C₆H₆BrNO
Molecular Weight: 188,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16999374
CAS-Nummer: 100367-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR