4-Iodo-2-methoxyphenol
Lieferant: Apollo Scientific
|
Gefahr
|
OR46089-1G
OR46089-25G
OR46089-5G
APOSOR46089-1GEA
56.7
EUR
APOSOR46089-1G
APOSOR46089-25G
APOSOR46089-5G
4-Iodo-2-methoxyphenol
4-Iodo-2-methoxyphenol
Formel:
C₇H₇IO₂ Schmelzpunkt: 37…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061924 CAS-Nummer: 203861-62-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...