Methyl 3-formyl-2-hydroxybenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR46142-1G
OR46142-250MG
APOSOR46142-1GEA
640
EUR
APOSOR46142-1G
APOSOR46142-250MG
Methyl 3-formyl-2-hydroxybenzoate 95%
Methyl 3-formyl-2-hydroxybenzoate
Formel:
C₉H₈O₄ Molecular Weight: 180,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01067661 CAS-Nummer: 3775-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...