5-Bromo-2-chloroiodobenzene 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-chloro-2-iodobenzene

OR46215-25G OR46215-5G OR46215-100G
APOSOR46215-25GEA 87 EUR
APOSOR46215-25G APOSOR46215-5G APOSOR46215-100G
5-Bromo-2-chloroiodobenzene 95+%
5-Bromo-2-chloroiodobenzene
Formel: C₆H₃BrClI
Molecular Weight: 317.349 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07780669
CAS-Nummer: 774608-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR