3-Chloro-5-iodoanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-5-iodophenyl methyl ether, 1-Chloro-3-iodo-5-methoxybenzene
OR46237-1G
OR46237-250MG
APOSOR46237-1GEA
343
EUR
APOSOR46237-1G
APOSOR46237-250MG
3-Chloro-5-iodoanisole
3-Chloro-5-iodoanisole
Formel:
C₇H₆ClIO Molecular Weight: 268,479 g/mol Siedepunkt: 87…89 °C (1,5 mm Hg) Dichte: 1,8961 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393318 CAS-Nummer: 861800-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...