(+)-Corey lactone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3aS,4R,5S,6aR)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (-)-Corey Lactone Diol, (3aS,4R,5S,6aR)-5-Hydroxy-4-(hydroxymethyl)-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one

OR46256-5G
APOSOR46256-5GEA 134 EUR
APOSOR46256-5G
(+)-Corey lactone 95%
(+)-Corey lactone
Formel: C₈H₁₂O₄
Molecular Weight: 172,178 g/mol
Schmelzpunkt: 114…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00274285
CAS-Nummer: 76704-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR