Methyl 5-bromo-3-chloropyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-chloro-2-(methoxycarbonyl)pyridine, Methyl 5-bromo-3-chloropicolinate

OR46525-1G OR46525-250MG OR46525-5G OR46525-25G
APOSOR46525-1GEA 21.9 EUR
APOSOR46525-1G APOSOR46525-250MG APOSOR46525-5G APOSOR46525-25G
Methyl 5-bromo-3-chloropyridine-2-carboxylate
Methyl 5-bromo-3-chloropyridine-2-carboxylate
Formel: C₇H₅BrClNO₂
Molecular Weight: 250,477 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698107
CAS-Nummer: 1214336-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR