Methyl 4-(bromoacetyl)benzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Methoxycarbonyl)phenacyl bromide, Methyl 4-(2-bromoacetyl)benzoate, 2-Bromo-4'-(methoxycarbonyl)acetophenone
OR46535-1G
OR46535-250MG
APOSOR46535-1GEA
72.5
EUR
APOSOR46535-1G
APOSOR46535-250MG
Methyl 4-(bromoacetyl)benzoate
Methyl 4-(bromoacetyl)benzoate
Formel:
C₁₀H₉BrO₃ Molecular Weight: 257,081 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08062347 CAS-Nummer: 56893-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...