4-Bromo-2-iodobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-2-iodophenyl)methanol

OR46536-1G OR46536-250MG OR46536-5G
APOSOR46536-1GEA 178 EUR
APOSOR46536-1G APOSOR46536-250MG APOSOR46536-5G
4-Bromo-2-iodobenzyl alcohol
4-Bromo-2-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391656
CAS-Nummer: 1261438-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR