4-Amino-3-bromo-5-nitrobenzonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-cyano-6-nitroaniline, 2-Amino-3-bromo-5-cyanonitrobenzene
OR46543-1G
OR46543-5G
OR46543-10G
APOSOR46543-1GEA
62.5
EUR
APOSOR46543-1G
APOSOR46543-5G
APOSOR46543-10G
4-Amino-3-bromo-5-nitrobenzonitrile 95%
4-Amino-3-bromo-5-nitrobenzonitrile
Formel:
C₇H₄BrN₃O₂ Molecular Weight: 242,03 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22383659 CAS-Nummer: 1239720-33-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...