3-Bromo-5-nitrobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3-(bromomethyl)-5-nitrobenzene, 3-Bromo-5-(bromomethyl)nitrobenzene
OR46553-1G
OR46553-250MG
OR46553-5G
APOSOR46553-1GEA
325
EUR
APOSOR46553-1G
APOSOR46553-250MG
APOSOR46553-5G
3-Bromo-5-nitrobenzyl bromide
3-Bromo-5-nitrobenzyl bromide
Formel:
C₇H₅Br₂NO₂ Molecular Weight: 294,928 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040230 CAS-Nummer: 139194-80-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...