5-Bromo-2-iodobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-2-iodophenyl)methanol

OR46556-1G OR46556-250MG OR46556-5G
APOSOR46556-1GEA 27.7 EUR
APOSOR46556-1G APOSOR46556-250MG APOSOR46556-5G
5-Bromo-2-iodobenzyl alcohol
5-Bromo-2-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09745850
CAS-Nummer: 199786-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR