5-Methoxy-1H-indazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR46578-1G OR46578-5G
APOSOR46578-1GEA 203 EUR
APOSOR46578-1G APOSOR46578-5G
5-Methoxy-1H-indazole-3-carboxylic acid 95%
5-Methoxy-1H-indazole-3-carboxylic acid
Formel: C₉H₈N₂O₃
Molecular Weight: 192.171 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03840712
CAS-Nummer: 90417-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR