3-Bromo-4,5-dichloropyridine 97%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dichloro-5-bromopyridine

OR46598-250MG OR46598-1G OR46598-5G
APOSOR46598-250MGEA 63.8 EUR
APOSOR46598-250MG APOSOR46598-1G APOSOR46598-5G
3-Bromo-4,5-dichloropyridine 97%
3-Bromo-4,5-dichloropyridine
Formel: C₅H₂BrCl₂N
Molecular Weight: 226,886 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014545
CAS-Nummer: 1001056-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR