N-Methylpropane-1,3-diamine, N-BOC protected hydrochloride 97%
Lieferant: Apollo Scientific
Achtung
Synonyme: tert-Butyl (3-aminopropyl)(methyl)carbamate hydrochloride, N-(tert-Butoxycarbonyl)-N-methylpropane-1,3-diamine hydrochloride, tert-Butyl (3-aminoprop-1-yl)methylcarbamate hydrochloride
OR46601-1G OR46601-5G
APOSOR46601-1GEA 85.7 EUR
APOSOR46601-1G APOSOR46601-5G
N-Methylpropane-1,3-diamine, N-BOC protected hydrochloride 97%
N-Methylpropane-1,3-diamine, N-BOC protected hydrochloride
Formel: C₉H₂₀N₂O₂·ClH
Molecular Weight: 224.728 g/mol
Schmelzpunkt: 138…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04973126
CAS-Nummer: 1188263-67-3