Pyridine-3-sulphonamide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Sulphamoylpyridine, 3-(Aminosulphonyl)pyridine

OR46603-1G OR46603-5G
APOSOR46603-1GEA 63.8 EUR
APOSOR46603-1G APOSOR46603-5G
Pyridine-3-sulphonamide 98%
Pyridine-3-sulphonamide
Formel: C₅H₆N₂O₂S
Molecular Weight: 158,178 g/mol
Schmelzpunkt: 106…111 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD08690712
CAS-Nummer: 2922-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR