2-(4-Pyridinyloxy)-ethanamine hydrochloride

Lieferant: Apollo Scientific

OR46625-1G OR46625-250MG OR46625-500MG OR46625-2.5G OR46625-5G
APOSOR46625-1GEA 432 EUR
APOSOR46625-1G APOSOR46625-250MG APOSOR46625-500MG APOSOR46625-2.5G APOSOR46625-5G
2-(4-Pyridinyloxy)-ethanamine hydrochloride
2-(4-Pyridinyloxy)-ethanamine hydrochloride
Formel: C₇H₁₀N₂O·ClH
Molecular Weight: 174,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28100442
CAS-Nummer: 1992956-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR