Cyclooctanecarbaldehyde

Lieferant: Apollo Scientific

OR46637-1G OR46637-250MG OR46637-500MG
APOSOR46637-1GEA 452 EUR
APOSOR46637-1G APOSOR46637-250MG APOSOR46637-500MG
Cyclooctanecarbaldehyde
Cyclooctanecarbaldehyde
Formel: C₉H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00001747
CAS-Nummer: 6688-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR