Phenylpyrrolidin-1-ylacetic acid hydrochloride

Lieferant: Apollo Scientific

OR46654-1G OR46654-50MG OR46654-250MG OR46654-500MG
APOSOR46654-1GEA 517 EUR
APOSOR46654-1G APOSOR46654-50MG APOSOR46654-250MG APOSOR46654-500MG
Phenylpyrrolidin-1-ylacetic acid hydrochloride
Phenylpyrrolidin-1-ylacetic acid hydrochloride
Formel: C₁₂H₁₅NO₂·ClH
Molecular Weight: 241,714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801210
CAS-Nummer: 100390-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR