4-chloro-carbonyl piperidine hydrochloride
Lieferant: Apollo Scientific
OR46688-1G
OR46688-250MG
OR46688-500MG
OR46688-2.5G
APOSOR46688-1GEA
510
EUR
APOSOR46688-1G
APOSOR46688-250MG
APOSOR46688-500MG
APOSOR46688-2.5G
4-chloro-carbonyl piperidine hydrochloride
4-chloro-carbonyl piperidine hydrochloride
Formel:
C₆H₁₀ClNO·ClH Molecular Weight: 184.064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26099148 CAS-Nummer: 42060-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...