1-(3-Chloro-4-methoxyphenyl)propan-1-one
Lieferant: Apollo Scientific
OR46705-1G
OR46705-50MG
OR46705-250MG
OR46705-500MG
APOSOR46705-1GEA
440
EUR
APOSOR46705-1G
APOSOR46705-50MG
APOSOR46705-250MG
APOSOR46705-500MG
1-(3-Chloro-4-methoxyphenyl)propan-1-one
1-(3-Chloro-4-methoxyphenyl)propan-1-one
Formel:
C₁₀H₁₁ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11199128 CAS-Nummer: 4394-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...