2-Bromo-3-cyclopropylthiophene
Lieferant: Apollo Scientific
|
Achtung
|
OR46744-1G
OR46744-250MG
APOSOR46744-1GEA
72.5
EUR
APOSOR46744-1G
APOSOR46744-250MG
2-Bromo-3-cyclopropylthiophene
2-Bromo-3-cyclopropylthiophene
Formel:
C₇H₇BrS Molecular Weight: 203.099 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16618635 CAS-Nummer: 29481-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...