6-Chloro-3-iodopyridin-2-amine 98%

Lieferant: Apollo Scientific
Achtung

OR46764-5G
APOSOR46764-5GEA 108 EUR
APOSOR46764-5G
6-Chloro-3-iodopyridin-2-amine 98%
6-Chloro-3-iodopyridin-2-amine
Formel: C₅H₄ClIN₂
Molecular Weight: 254,456 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13181515
CAS-Nummer: 800402-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR