2-Amino-6-bromoindole hydrochloride 95%

Lieferant: Apollo Scientific

OR46769-1G OR46769-250MG OR46769-5G
APOSOR46769-1GEA 551 EUR
APOSOR46769-1G APOSOR46769-250MG APOSOR46769-5G
2-Amino-6-bromoindole hydrochloride 95%
2-Amino-6-bromoindole hydrochloride
Formel: C₈H₇BrN₂·ClH
Molecular Weight: 247,52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640237
CAS-Nummer: 28493-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR