2-Carbamoyl-6-nitrobenzoic acid
Lieferant: Apollo Scientific
OR470009-25G
OR470009-5G
APOSOR470009-25GEA
601
EUR
APOSOR470009-25G
APOSOR470009-5G
2-Carbamoyl-6-nitrobenzoic acid
2-Carbamoyl-6-nitrobenzoic acid
Formel:
C₈H₆N₂O₅ Molecular Weight: 210.144 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00088952 CAS-Nummer: 65911-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...