D-N-Benzylserine
Lieferant: Apollo Scientific
Synonyme:
(R)-2-(Benzylamino)-3-hydroxypropanoic acid, N-D-Bzl-Ser-OH
OR470024-5G
APOSOR470024-5GEA
58
EUR
APOSOR470024-5G
D-N-Benzylserine
D-N-Benzylserine
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198844 CAS-Nummer: 106910-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...