2-(Cyclohexyloxy)ethanol

Lieferant: Apollo Scientific

OR470058-25G OR470058-5G
APOSOR470058-25GEA 916 EUR
APOSOR470058-25G APOSOR470058-5G
2-(Cyclohexyloxy)ethanol
2-(Cyclohexyloxy)ethanol
Formel: C₈H₁₆O₂
Molecular Weight: 144,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134514
CAS-Nummer: 1817-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR