3-[4-(Methylthio)phenoxy]propylamine

Lieferant: Apollo Scientific

OR470095-1G OR470095-5G
APOSOR470095-1GEA 306 EUR
APOSOR470095-1G APOSOR470095-5G
3-[4-(Methylthio)phenoxy]propylamine
3-[4-(Methylthio)phenoxy]propylamine
Formel: C₁₀H₁₅NOS
Molecular Weight: 197,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16495893
CAS-Nummer: 1226776-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR