3-[2-(Dimethylamino)ethoxy]-4-methoxybenzonitrile
Lieferant: Apollo Scientific
OR470108-1G
OR470108-5G
APOSOR470108-1GEA
306
EUR
APOSOR470108-1G
APOSOR470108-5G
3-[2-(Dimethylamino)ethoxy]-4-methoxybenzonitrile
3-[2-(Dimethylamino)ethoxy]-4-methoxybenzonitrile
Formel:
C₁₂H₁₆N₂O₂ Molecular Weight: 220,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09711359 CAS-Nummer: 938239-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...