tert-Butyl (S)-4-(Cbz-amino)-5-methyl-3-oxohexanoate

Lieferant: Apollo Scientific

OR470109-1G OR470109-5G
APOSOR470109-1GEA 0 EUR
APOSOR470109-1G APOSOR470109-5G
tert-Butyl (S)-4-(Cbz-amino)-5-methyl-3-oxohexanoate
tert-Butyl (S)-4-(Cbz-amino)-5-methyl-3-oxohexanoate
Formel: C₁₉H₂₇NO₅
Molecular Weight: 349.421 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16495901
CAS-Nummer: 191731-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR