2-(Methyl-2-pyridylamino)acetic acid

Lieferant: Apollo Scientific

OR470122-1G OR470122-5G
APOSOR470122-1GEA 397 EUR
APOSOR470122-1G APOSOR470122-5G
2-(Methyl-2-pyridylamino)acetic acid
2-(Methyl-2-pyridylamino)acetic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09932969
CAS-Nummer: 1016519-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR