4-Methoxybenzo[d]oxazole-2(3H)-thione

Lieferant: Apollo Scientific

OR470124-1G OR470124-5G
APOSOR470124-1GEA 108 EUR
APOSOR470124-1G APOSOR470124-5G
4-Methoxybenzo[d]oxazole-2(3H)-thione
4-Methoxybenzo[d]oxazole-2(3H)-thione
Formel: C₈H₇NO₂S
Molecular Weight: 181.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17171374
CAS-Nummer: 1246471-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR