Methyl N-Boc-3-aminophenylacetate

Lieferant: Apollo Scientific

OR470146-1G OR470146-250MG
APOSOR470146-1GEA 499 EUR
APOSOR470146-1G APOSOR470146-250MG
Methyl N-Boc-3-aminophenylacetate
Methyl N-Boc-3-aminophenylacetate
Formel: C₁₄H₁₉NO₄
Molecular Weight: 265,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071010
CAS-Nummer: 1256633-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR