2-[2-(Boc-amino)ethoxy]benzamide
Lieferant: Apollo Scientific
OR470158-1G
OR470158-5G
APOSOR470158-1GEA
306
EUR
APOSOR470158-1G
APOSOR470158-5G
2-[2-(Boc-amino)ethoxy]benzamide
2-[2-(Boc-amino)ethoxy]benzamide
Formel:
C₁₄H₂₀N₂O₄ Molecular Weight: 280,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071034 CAS-Nummer: 1256633-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...