4-[2-(Boc-amino)ethoxy]benzamide

Lieferant: Apollo Scientific

OR470159-1G OR470159-5G
APOSOR470159-1GEA 306 EUR
APOSOR470159-1G APOSOR470159-5G
4-[2-(Boc-amino)ethoxy]benzamide
4-[2-(Boc-amino)ethoxy]benzamide
Formel: C₁₄H₂₀N₂O₄
Molecular Weight: 280,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071035
CAS-Nummer: 1256633-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR